Learn More
Check All Valve SCV-150-SS-UE-1/8
SDP
Catalog No. NC0493847
Encompass
1-1/2 in. SCV style check valve made out 316SS body material with a USP class VI EPDM oring seat and a 1/8 psi spring cracking pressure which is made out of 316SS. The outside oring is also USP class VI EPDM.
Catalog No. NC0493847 Supplier Check All Valve Supplier No. SCJSSUE.125SSO
1-1/2 in. SCV style check valve made out 316SS body material with a USP class VI EPDM oring seat and a 1/8 psi spring cracking pressure which is made out of 316SS. The outside oring is also USP class VI EPDM.