Learn More
Check All Valve SCV-300-SS-UE-1/2
SDP
Catalog No. NC0493845
Encompass
3 in. SCV style check valve made out 316SS body material with a USP class VI EPDM oring seat and a 1/2 psi spring cracking pressure which is also made out 316SS. The outside oring is also USP class VI EPDM.
Catalog No. NC0493845 Supplier Check All Valve Supplier No. SCMSSUE.500SSO
3 in. SCV style check valve made out 316SS body material with a USP class VI EPDM oring seat and a 1/2 psi spring cracking pressure which is also made out 316SS. The outside oring is also USP class VI EPDM.