Learn More
Waters Corp Waters BioResolve SCX mAb Column, 3 μm, 2.1 mm x 50 mm
Catalog No. NC1684442 Shop All Waters Corporation Products
Encompass_Preferred
This BioResolve SCX mAb Column, 3 μm, 2.1 mm x 50 mm, is corrosion-resistant with non-porous, polymer-based particles. It offers reproducible, high-resolution, charge-based separations for monoclonal antibodies (mAbs) and proteins in LC and LC-MS applications, using robust salt or pH gradient methods. QC testing with a mAb Charge Variant Standard ensures consistent performance.
  • Designed for charge variant analysis.
  • Withstands extreme chromatographic conditions.
  • Robust salt or pH gradient methods for high-resolution mAb/fragment analysis.
  • Non-porous polymeric particles with negatively charged sulfonic acid ligands.
  • Compatible with UHPLC, HPLC, and UPLC systems.
  • Includes a mAb Charge Variant Standard for proper LC-MS system operation.
  • Combines with CX pH Buffer Concentrates for enhanced performance.
  • Uses SCX mAb VanGuard FIT Cartridge for enhanced protection.
Catalog No. NC1684442 Supplier Waters Corp Supplier No. 186009054
This BioResolve SCX mAb Column, 3 μm, 2.1 mm x 50 mm, is corrosion-resistant with non-porous, polymer-based particles. It offers reproducible, high-resolution, charge-based separations for monoclonal antibodies (mAbs) and proteins in LC and LC-MS applications, using robust salt or pH gradient methods. QC testing with a mAb Charge Variant Standard ensures consistent performance.
  • Designed for charge variant analysis.
  • Withstands extreme chromatographic conditions.
  • Robust salt or pH gradient methods for high-resolution mAb/fragment analysis.
  • Non-porous polymeric particles with negatively charged sulfonic acid ligands.
  • Compatible with UHPLC, HPLC, and UPLC systems.
  • Includes a mAb Charge Variant Standard for proper LC-MS system operation.
  • Combines with CX pH Buffer Concentrates for enhanced performance.
  • Uses SCX mAb VanGuard FIT Cartridge for enhanced protection.