Learn More
NANOMICRO TECHNOLOGIES INC BioCore SCX LC Analytical Column, 10um, 4.6x250 mm, PEEK
Catalog No. NC2814234
Encompass_Preferred
BioCore SCX is a family of high-performance, strong cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic sulfonic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2814234 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B41110000004625P
BioCore SCX is a family of high-performance, strong cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic sulfonic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.