Learn More
Analytical Sales & Services SCX SPE 96-Well Plate, 1.0mL/100mg Bed
Catalog No. 5000807011
Encompass
Silica Based Strong Cation Exchange SPE, 1mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-07011 Supplier Analytical Sales & Services Supplier No. 9610SCX100
Silica Based Strong Cation Exchange SPE, 1mL 96 Well Filter Plate, 100mg per well