Learn More
Analytical Sales & Services SCX SPE 96-Well Plate, 1.0mL/25mg Bed
Catalog No. 5000806357
Encompass
Silica Based Strong Cation Exchange SPE, 1mL 96 Well Filter Plate, 25mg per well
Catalog No. 50-008-06357 Supplier Analytical Sales & Services Supplier No. 9610SCX25
Add to Cart
Add to Cart
Silica Based Strong Cation Exchange SPE, 1mL 96 Well Filter Plate, 25mg per well