Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore SCX LC Preparative Column, 5 m, 21.2250 mm, Stainless Steel
Catalog No. NC2815228
Encompass_Preferred
BioCore SCX is a family of high-performance, strong cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic sulfonic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2815228 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B41105000021225S
Add to Cart
Add to Cart
Request a Quote
BioCore SCX is a family of high-performance, strong cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic sulfonic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.