Learn More
Waters Corp SCX Start Up Kit, BioResolve SCX mAb, 4.6 mm, 50 mm
Catalog No. 502390452 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The SCX Start Up Kit is an all-in-one solution designed for monoclonal antibody (mAb) assays, offering reproducible, high-resolution, charged-based separations of mAbs and other proteins in LC and LC-MS applications. It utilizes corrosion-resistant columns with 3 μm non-porous, polymer-based particles and negatively charged sulfonic acid ligands. The kit includes a mAb Charge Variant Standard, pH Concentrates, and certified containers.
  • Reproducible, high-resolution, charged-based separations of mAbs and other proteins.
  • Compatible with LC and LC-MS applications.
  • Uses robust salt or pH gradient methods.
  • Includes a mAb Charge Variant Standard for system performance benchmarking.
  • Includes pH Concentrates A and B for charge variant profiling.
  • LC-MS system agnostic, compatible with UHPLC, UPLC, and HPLC systems.
  • Designed for column protection.
  • Includes an eCord.
Catalog No. 50-239-0452 Supplier Waters Corp Supplier No. 176004357
The SCX Start Up Kit is an all-in-one solution designed for monoclonal antibody (mAb) assays, offering reproducible, high-resolution, charged-based separations of mAbs and other proteins in LC and LC-MS applications. It utilizes corrosion-resistant columns with 3 μm non-porous, polymer-based particles and negatively charged sulfonic acid ligands. The kit includes a mAb Charge Variant Standard, pH Concentrates, and certified containers.
  • Reproducible, high-resolution, charged-based separations of mAbs and other proteins.
  • Compatible with LC and LC-MS applications.
  • Uses robust salt or pH gradient methods.
  • Includes a mAb Charge Variant Standard for system performance benchmarking.
  • Includes pH Concentrates A and B for charge variant profiling.
  • LC-MS system agnostic, compatible with UHPLC, UPLC, and HPLC systems.
  • Designed for column protection.
  • Includes an eCord.