Learn More
Waters Corp SCX Start Up Kit, BioResolve SCX mAb Column, 2.1 mm x 50 mm, VanGuard FIT
Catalog No. 502392892 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters SCX Start Up Kit is designed for strong-cation exchange chromatographic techniques used in mAb analyses. It includes BioResolve SCX mAb Columns, VanGuard FIT, pH concentrates, and a mAb Standard to streamline the workflow for understanding mAb-based biotherapeutic drugs.
  • Designed for reliable and accurate charge variant quantitation of mAbs using optical or LC-MS compatible mobile phases.
  • Utilizes non-porous, polymeric particles for optimal protein diffusion kinetics and pressure tolerance across UPLC, UHPLC, and HPLC systems.
  • Includes pH concentrates A (pH 5.0) and B (pH 10.2) to facilitate charge variant profiling.
  • Protects columns from contamination and variability, integrating into existing analytical workflows.
  • Certified LDPE containers are included for the concentrates.
  • mAb charge variant standard for system performance verification.
Catalog No. 50-239-2892 Supplier Waters Corp Supplier No. 176004351
The Waters SCX Start Up Kit is designed for strong-cation exchange chromatographic techniques used in mAb analyses. It includes BioResolve SCX mAb Columns, VanGuard FIT, pH concentrates, and a mAb Standard to streamline the workflow for understanding mAb-based biotherapeutic drugs.
  • Designed for reliable and accurate charge variant quantitation of mAbs using optical or LC-MS compatible mobile phases.
  • Utilizes non-porous, polymeric particles for optimal protein diffusion kinetics and pressure tolerance across UPLC, UHPLC, and HPLC systems.
  • Includes pH concentrates A (pH 5.0) and B (pH 10.2) to facilitate charge variant profiling.
  • Protects columns from contamination and variability, integrating into existing analytical workflows.
  • Certified LDPE containers are included for the concentrates.
  • mAb charge variant standard for system performance verification.