Learn More
Waters Corp SCX Start Up Kit, BioResolve SCX mAb Column, 4.6 mm x 100 mm, VanGuard FIT
Catalog No. 502392912 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This workflow kit streamlines monoclonal antibody quantitation via strong cation-exchange chromatography. It includes corrosion-resistant SCX mAb columns with non-porous, polymer-based particles and negatively charged sulfonic acid ligands for high-resolution LC and LC-MS separations. Also features VanGuard™ FIT for extended column life, a mAb Charge Variant Standard, and pH concentrates.
  • All-in-one kit for accurate mAb quantitation
  • SCX mAb columns provide high-resolution, charge-based separations
  • Columns use non-porous particles and sulfonic acid ligands
  • Wide pH range for diverse analytes
  • VanGuard™ FIT cartridge protects columns, extending lifespan
  • Kit includes two columns, pH concentrates, and a mAb standard
Catalog No. 50-239-2912 Supplier Waters Corp Supplier No. 176004354
This workflow kit streamlines monoclonal antibody quantitation via strong cation-exchange chromatography. It includes corrosion-resistant SCX mAb columns with non-porous, polymer-based particles and negatively charged sulfonic acid ligands for high-resolution LC and LC-MS separations. Also features VanGuard™ FIT for extended column life, a mAb Charge Variant Standard, and pH concentrates.
  • All-in-one kit for accurate mAb quantitation
  • SCX mAb columns provide high-resolution, charge-based separations
  • Columns use non-porous particles and sulfonic acid ligands
  • Wide pH range for diverse analytes
  • VanGuard™ FIT cartridge protects columns, extending lifespan
  • Kit includes two columns, pH concentrates, and a mAb standard