Learn More
Aviva Systems Biology Corp SCYE1 Antibody - N-terminal region (ARP40551_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40551_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCYE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SCYE1. Alternate names: p43, HLD3, EMAP2, SCYE1, EMAPIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57518