Learn More
Aviva Systems Biology Corp SCYE1 Antibody - middle region (ARP40552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCYE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SCYE1. Alternate names: AIMP1, EMAP II, EMAP-2, EMAP-II, EMAP2, EMAPII, p43, HLD3, SCYE1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57519