Learn More
Aviva Systems Biology Corp SCYL1 Antibody - C-terminal region (ARP61984_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61984_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCYL1. It was validated on Western Blot. . Alternate names: GKLP, HT019, MGC78454, NKTL, NTKL, P105, TAPK, TEIF, TRAPApplicationWB. Species reactivity: Horse, Human, Mouse, Rat
Catalog No. 50-287-531