Learn More
Cayman Chemical scyllo-Inositol hexakisphosphate (sodium salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502894199
Encompass_Preferred
A stereoisomer of IP6; identified in soil samples from Madagascar, New Zealand, and the Falkland Islands and in freshwater sediment from Danish lakes; has been used as a negative control for the stereospecific effects of myo-inositol hexakisphosphate on IK,in in intact guard cells
Catalog No. 50-289-4199 Supplier Cayman Chemical Supplier No. 90023395 MG
A stereoisomer of IP6; identified in soil samples from Madagascar, New Zealand, and the Falkland Islands and in freshwater sediment from Danish lakes; has been used as a negative control for the stereospecific effects of myo-inositol hexakisphosphate on IK,in in intact guard cells