Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

scyllo-Inositol 99.0+%, TCI America™

Catalog No. I06311G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06311G Supplier Sigma Organic Chemistry Supplier No. I06311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 488-59-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00077932,MFCD00065455,MFCD00003863,MFCD00272608,MFCD00799555,MFCD00799556,MFCD01321249
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES OC1C(O)C(O)C(O)C(O)C1O

Specifications

Melting Point 350°C
Color White
Quantity 1 g
Formula Weight 180.16
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material scyllo-Inositol
EINECSNumber (3)-3265
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.