Learn More
Medchemexpress LLC HY-11087 10mg Medchemexpress, SD 0006 CAS:271576-80-8 Purity:>98%
SDP
Catalog No. 501968808
Encompass_Preferred
Medchemexpress, HY-11087 10mg SD 0006 CAS:271576-80-8 SD 0006 (SD-06) is an orally active, selective, ATP-competitive and potent diaryl pyrazole inhibitor of p38α MAP kinase, with an IC50 of 110 nM for p38α. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8808 Supplier Medchemexpress LLC Supplier No. HY1108710MG
Medchemexpress, HY-11087 10mg SD 0006 CAS:271576-80-8 SD 0006 (SD-06) is an orally active, selective, ATP-competitive and potent diaryl pyrazole inhibitor of p38α MAP kinase, with an IC50 of 110 nM for p38α. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.