Learn More
Enzo Life Sciences SD-169 (10mg). CAS: 1670-87-7
SDP
Supplier:  Enzo Life Sciences BMLEI3710010
Encompass_Preferred
A selective inhibitor of p38α (IC50 = 3.2nM) and p38β (IC50 = 3.2nM), but not p38γ (IC50 > 50μM), ERK2, JNK1 or MAPKAP2. Lowers blood glucose and improves glucose homeostasis in nonobese diabetic mice. Enhances axonal regeneration in animals with nerve transection. Alternative name: 1H-Indole-5-carboxamide. Purity: ≥96.5% (HPLC). Solubility: Soluble in water (1.5mg/ml); DMSO (15mg/ml) or Ethanol (15mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0756