Learn More
Medchemexpress LLC HY-W015445 50mg Medchemexpress, SD-169 CAS:1670-87-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01544550MG
Encompass_Preferred
Medchemexpress, HY-W015445 50mg SD-169 CAS:1670-87-7 SD-169 is an orally active ATP-competitive inhibitor of p38α MAPK, with an IC50 of 3.2 nM. SD-169 also weakly inhibits p38β MAPK with an IC50 of 122 nM. SD-169 prevents the development and progression of diabetes by inhibiting T cell infiltration and activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3377