Learn More
Selleck Chemical LLC SD-208
SDP
Catalog No. 5000000289
Encompass_Preferred
SD-208 is a selective TGF- RI (ALK5) inhibitor with IC50 of 48 nM 100-fold selectivity over TGF- RII
Catalog No. 50-000-00289 Supplier Selleck Chemical LLC Supplier No. S7624100MG
Add to Cart
Add to Cart
SD-208 is a selective TGF- RI (ALK5) inhibitor with IC50 of 48 nM 100-fold selectivity over TGF- RII