Learn More
Selleck Chemical LLC SD-208 5mg 627536-09-8
SDP
Supplier:  Selleck Chemical LLC S76245MG
Encompass_Preferred
SD-208 is a selective TGF-?RI (ALK5) inhibitor with IC50 of 48 nM, >100-fold selectivity over TGF-?RII. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1322747