Learn More
Boca Scientific Inc SD Polymerase, 10 U/ul, 200 U
Supplier:  Boca Scientific Inc 108702
Encompass
SD Polymerase is a novel, artificial polymerase with a strong strand displacement activity and heat stability up to 93°C. With SD Polymerase, the possibilities of isothermal amplification and regular PCR can be combined. The high heat stability of SD Polymerase enhances the efficiency of isothermal amplification by initial denaturation steps to increase the accessibility of single strands. The initial heating-up improves the specificity in applications such as LAMP, library generation, PCDR, WGA, standard PCR, and genome sequencing. The special properties of SD Polymerase provide greater yields, speed and efficiency than Taq Polymerase. SD Polymerase is suitable for high GC-content and difficult targets. Long Range PCR can be performed with target sizes up to 30 kb. SD Polymerase has 5’-3’ polymerase and 5’-3’ strand displacement activities, as well as 3’-A-overhangs. It does not have any exonuclease activity.
Catalog No. 50-168-5638