Learn More
iQ Biosciences SD Rat Bone Marrow Cells, 10M cells/vial
Catalog No. NC2855602
Encompass_Preferred
High-quality, high-viability (typically > 70%) bone marrow cells from the Sprague Dawley, rat a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2855602 Supplier iQ Biosciences Supplier No. IQBRBM101
High-quality, high-viability (typically > 70%) bone marrow cells from the Sprague Dawley, rat a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.