Learn More
iQ Biosciences SD Rat Splenocytes, 50M cells/vial
Catalog No. NC2384411
Encompass_Preferred
High-quality, high-viability (typically > 70%) splenocytes from the Sprague Dawley rat, a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2384411 Supplier iQ Biosciences Supplier No. IQBRSP102
High-quality, high-viability (typically > 70%) splenocytes from the Sprague Dawley rat, a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.