Learn More
Description
- A three-colour LED lighting system - blue, green and red - and new display icons make the tester easy to use and intuitive to operate
- The use of energy saving LEDs and 4 x AAA batteries greatly extends the life of the new Pocket Tester range to up to 2000 hours
- With the included lanyard, each SD Series tester can be worn comfortably around the neck or attached to a pocket, waistband or belt with the clip on the head of the tester
- Comes with: Case, batteries, lanyard, 10x pH 4, 7, 10 buffer tablets, manual, warranty guide, certificate of compliance
Specifications
Specifications
| Includes | Case, Lanyard, Buffer Set, Storage Solution |
| Accuracy (Temperature) | 0.5°C |
| Accuracy (pH) | 0.01 |
| Measurement Modes | pH |
| Calibration | pH |
| Range (pH) | 0 to 14 |
| Resolution (pH) | 0.01 |
| Range (mV) | -400 to 400 mV |
| Calibration Points | 1 to 3 |
| Temperature Range (Metric) | 0°C to 50°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.