Learn More
Penn Valley Chemical SD-20 All Purpose Cleaner, 55 gallon
Catalog No. NC0390437
Encompass_Preferred
Formulated without soap, free alkali, or ammonia, SD-20 is an all-surface cleaner and may be used on floors, painted walls, woodwork, marble, plastic, metal and other hard surfaces impervious to water.
Catalog No. NC0390437 Supplier Penn Valley Chemical Supplier No. SPSD2055
Formulated without soap, free alkali, or ammonia, SD-20 is an all-surface cleaner and may be used on floors, painted walls, woodwork, marble, plastic, metal and other hard surfaces impervious to water.