Description
- A three-colour LED lighting system - blue, green and red - and new display icons make the tester easy to use and intuitive to operate
- The use of energy saving LEDs and 4 x AAA batteries greatly extends the life of the new pocket tester range to up to 2000 hours
- With the included lanyard, each SD Series tester can be worn comfortably around the neck or attached to a pocket, waistband or belt with the clip on the head of the tester
- Comes with: Device, Case, batteries, lanyard, 3M KCl Storage Solution, Multilingual Manual, Warranty Declaration, Certificate of Compliance
Specifications
Specifications
| Type | ORP Pen Meter with Temperature Readout |