Learn More
Description
- Rugged, water resistant (IP 67)
- PC interface (USB / serial or analog)
- Automatic buffer detection
- Data logger and alarm function (min./max
- The protective housing not only guarantees safe working at all times, even in extreme environmental conditions. The stand attached to the back of the housing can also be used as a hanger or belt clip.
Specifications
Specifications
| Channels | Measurement: 2, Temperature: 2 |
| Display Type | Backlit Graphic LCD |
| IP Rating | IP67 |
| Measurement Modes | pH with mV and Temperature |
| Range (pH) | -2 to 16 |
| Resolution (pH) | 0.01 |
| Temperature Range (Metric) | -5 °C to +150°C |
| Product Type | pH/ORP Meter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.