Learn More
Description
- Intuitive user interface with graphical LCD-display
- Galvanically isolated measuring inputs
- Data transfer via Micro-USB in CSV format
- Robust protective armouring with two electrode holders
- Comes with: Instrument, Case, pH/temperature electrode type 231, Conductivity Cell LC12, pH buffer set (4, 7, 10), Storage Solution for pH/ ORP, COnductivity Solution (1413 μS/cm), NiMH battery pack, Micro USB cable, Manual, Warranty guide
Specifications
Specifications
| Channels | pH, Conductivity |
| Display Type | Backlit Graphic LCD |
| IP Rating | IP67 |
| Measurement Modes | pH |
| Range (Conductivity) | 8 to 200 mS/cm |
| Range (pH) | -2 to 16 |
| Range (TDS) | 0 to 200 g/L |
| Resolution (pH) | 0.01 |
| Temperature Range (Metric) | -10°C to +110°C |
| Product Type | pH/ Conductivity Meter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.