Learn More
Description
- Tests Conductivity, pH, Salinity, TDS, and Temperature
- A three-colour LED lighting system - blue, green and red - and new display icons make the tester easy to use and intuitive to operate
- The use of energy saving LEDs and 4 x AAA batteries greatly extends the life of the new Pocket Tester range to up to 2000 hours
- With the included lanyard, each SD Series tester can be worn comfortably around the neck or attached to a pocket, waistband or belt with the clip on the head of the tester
- pH buffer auto recognition: 1.68 / 4.00 / 7.00 / 10.01 / 12.45 pH
- Conductivity solution auto recognition: 84 μS/cm / 1413 μS/cm / 12 mS/cm
- Comes with: instrument, case, batteries, lanyard, 10x pH 4, 7, 10 buffer tablets, 3M KCl storage solution, manual, warranty guide, Certificate of Compliance
Specifications
Specifications
| Channels | Multiple |
| Display Type | Backlit Graphic LCD |
| IP Rating | IP67 |
| Measurement Modes | pH |
| Range (Conductivity) | 0.01 to 20.00 mS/cm |
| Range (pH) | 0 to 14 |
| Range (TDS) | 0.01 to 20.00 ppt |
| Resolution (pH) | 0.01 |
| Temperature Range (Metric) | 0°C to 60°C |
| Product Type | Multiparameter Pen tester |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.