Learn More
General Laboratory Products Sabouraud Dextrose Agar, Cultivates Fungi, Easy-Pour Narrow Mouth Glass Bottle, 200 ml
Catalog No. NC2684466
Encompass_Preferred
Sabouraud Dextrose Agar (SDA) is a ready-to-use culture medium from General Laboratory Products Inc. designed for effective isolation and cultivation of fungi and aciduric microorganisms. Packaged in easy-pour, narrow-mouth glass bottles, with a total of 9 bottles per case, it ensures a reliable supply for laboratory needs. The low pH formulation inhibits most bacterial growth, distinguishing it from other media products.
Catalog No. NC2684466 Supplier General Laboratory Products Supplier No. 250EZ3200200
Sabouraud Dextrose Agar (SDA) is a ready-to-use culture medium from General Laboratory Products Inc. designed for effective isolation and cultivation of fungi and aciduric microorganisms. Packaged in easy-pour, narrow-mouth glass bottles, with a total of 9 bottles per case, it ensures a reliable supply for laboratory needs. The low pH formulation inhibits most bacterial growth, distinguishing it from other media products.