Learn More
Aat Bioquest SDA azide, 1mg
SDP
Catalog No. NC3930623
Encompass_Preferred
SDA azide contains both diazirine photoaffinity moiety and azide functional group. It is an excellent building block to introduce photoaffinity function into an alkyne-modified biomolecule via the well-known click chemistry. Diazirines are known for their ability to undergo photochemical reactions when exposed to ultraviolet (UV) light, specifically by forming highly reactive carbene intermediates that react with nearby molecules, forming covalent bonds. This property makes diazirines useful for studying protein-protein, protein-nucleic acid interactions, ligand-receptor binding, and other biomolecular interactions. It's important to note that diazirines are highly reactive and can be challenging to handle due to their instability. They require careful storage and handling, typically in a controlled environment.
Catalog No. NC3930623 Supplier Aat Bioquest Supplier No. 39004
SDA azide contains both diazirine photoaffinity moiety and azide functional group. It is an excellent building block to introduce photoaffinity function into an alkyne-modified biomolecule via the well-known click chemistry. Diazirines are known for their ability to undergo photochemical reactions when exposed to ultraviolet (UV) light, specifically by forming highly reactive carbene intermediates that react with nearby molecules, forming covalent bonds. This property makes diazirines useful for studying protein-protein, protein-nucleic acid interactions, ligand-receptor binding, and other biomolecular interactions. It's important to note that diazirines are highly reactive and can be challenging to handle due to their instability. They require careful storage and handling, typically in a controlled environment.