Learn More
General Laboratory Products Sabouraud Dextrose Agar (SDA) w/ Lecithin & Polysorbate 80, 250 mL, 9/cs
Catalog No. NC0831945
Encompass_Preferred
Sabouraud Dextrose Agar w/ Lecithin & Polysorbate 80 - 250ml, Easy-pour narrow mouth glass bottle, 9 per case. Base for the Cultivation of Fungi and Aciduric Microorganisms.
Catalog No. NC0831945 Supplier General Laboratory Products Supplier No. 250EZ7112250
Sabouraud Dextrose Agar w/ Lecithin & Polysorbate 80 - 250ml, Easy-pour narrow mouth glass bottle, 9 per case. Base for the Cultivation of Fungi and Aciduric Microorganisms.