Learn More
MilliporeSigma SDA w. LTHTh - ICR 30 mL, 120/pk
Catalog No. NC1633663
Encompass
Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media is used for environmental monitoring by determining the yeast and mold count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.
Catalog No. NC1633663 Supplier MilliporeSigma Supplier No. 1460050120
Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media is used for environmental monitoring by determining the yeast and mold count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.