Learn More
Cayman Chemical SDB-006 N-phenyl analog, Purity: ≥98%, CAS: 1430634-87-9, 10 mg
SDP
Catalog No. 502969053
Encompass_Preferred
A cannabimimetic indole that binds the CB1 and CB2 receptors with EC50 values of 21 and 140 nM, respectively; intended for research and forensic applications
Catalog No. 50-296-9053 Supplier Cayman Chemical Supplier No. 1604410 MG
A cannabimimetic indole that binds the CB1 and CB2 receptors with EC50 values of 21 and 140 nM, respectively; intended for research and forensic applications