Learn More
VEOLIA WTS SDBS Suitability Set
Catalog No. NC1236874
Encompass
Standard set to meet Japanese and Korean Pharmacopeia requirements for TOC analysis. This set contains (2) 40 mL glass vial standards: (1) Reagent water, and (1) 500 mg C/L as SDBS.
Catalog No. NC1236874 Supplier VEOLIA WTS Supplier No. STD 9003901
Standard set to meet Japanese and Korean Pharmacopeia requirements for TOC analysis. This set contains (2) 40 mL glass vial standards: (1) Reagent water, and (1) 500 mg C/L as SDBS.