Learn More
Aviva Systems Biology Corp Sdc3 Antibody - N-terminal region (ARP47052_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47052_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sdc3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MGC65603, MGC69616, Synd3, mKIAA0468, syn-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-285