Learn More
Teknova SDCAA Agar Plates with Tryptophan-40. 100mm, 20 plates/sleeve. Sterile. 4 pack.
SDP
Catalog No. 502387882
Encompass
Synthetic Defined Media (SDM) is a type of growth media that is formulated to provide cells or tissues with the nutrients they need to thrive. This media is used in research labs to study the effects of various chemicals on cells. SDM allows for more control over the environment in which cells are grown, and as a result, can provide insights that would be otherwise difficult to obtain.
Catalog No. 50-238-7882 Supplier Teknova Supplier No. S05514PK
Synthetic Defined Media (SDM) is a type of growth media that is formulated to provide cells or tissues with the nutrients they need to thrive. This media is used in research labs to study the effects of various chemicals on cells. SDM allows for more control over the environment in which cells are grown, and as a result, can provide insights that would be otherwise difficult to obtain.