Learn More
Aviva Systems Biology Corp SDCBP Antibody - middle region (ARP44535_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44535_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SDCBP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SDCBP. Alternate names: MDA-9, ST1, SYCL, TACIP18ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58835