Learn More
Aviva Systems Biology Corp SDCBP Antibody - middle region (ARP44537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SDCBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SDCBP. Alternate names: MDA-9, ST1, SYCL, TACIP18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58837