Learn More
Aviva Systems Biology Corp SDCBP2 Antibody - N-terminal region (ARP55326_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55326_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SDCBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SDCBP2. Alternate names: FLJ12256, SITAC18, ST-2, SITACApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-61469