Learn More
Cayman Chemical (S)-Desoxy-D2PM (hydrochloride), Purity: ≥98%, CAS: 188398-87-0, 10 mg
SDP
Catalog No. 502892772
Encompass_Preferred
Most commonly used as a chiral solvating agent for NMR analysis of chiral compounds; structurally related to desoxypipradrol, a dopamine transporter inhibitor and psychoactive stimulant
Catalog No. 50-289-2772 Supplier Cayman Chemical Supplier No. 900109510 MG
Most commonly used as a chiral solvating agent for NMR analysis of chiral compounds; structurally related to desoxypipradrol, a dopamine transporter inhibitor and psychoactive stimulant