Learn More
Medchemexpress LLC SDF-1 beta/CXCL12 H 2ug
SDP
Catalog No. 5000180530
Encompass_Preferred
SDF-1 beta (Stromal-derived factor-1 SDF-1 ) is a stromal derived CXC chemokine that signal through the CXCR4 receptor SDF-1 has chemotactic activity on B and T cells[1][2] SDF-1 beta/CXCL12 Protein Human (HEK293 Fc) is produced in HEK293 cells with a N-Terminal Fc-tag It consists of 72 amino acids (K22-M93)
Catalog No. 50-001-80530 Supplier Medchemexpress LLC Supplier No. HYP734112UG
SDF-1 beta (Stromal-derived factor-1 SDF-1 ) is a stromal derived CXC chemokine that signal through the CXCR4 receptor SDF-1 has chemotactic activity on B and T cells[1][2] SDF-1 beta/CXCL12 Protein Human (HEK293 Fc) is produced in HEK293 cells with a N-Terminal Fc-tag It consists of 72 amino acids (K22-M93)