Learn More
Medchemexpress LLC SDF-1 beta/CXCL12 H 2ug
SDP
Supplier:  Medchemexpress LLC HYP72872UG
Encompass_Preferred
SDF-1 beta (Stromal-derived factor-1 SDF-1 ) is a stromal derived CXC chemokine that signal through the CXCR4 receptor SDF-1 has chemotactic activity on B and T cells[1][2] SDF-1 beta/CXCL12 Protein Human (72a a) is produced in E coli and consists of 72 amino acids (K22-M93)
Catalog No. 50-001-81821