Learn More
Medchemexpress LLC HY-P7403 10ug Medchemexpress, SDF-1α/CXCL12 (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 502083160
Encompass_Preferred
Medchemexpress, HY-P7403 10ug SDF-1α/CXCL12 (CHO-expressed), Mouse CAS: Recombinant Mouse Stromal-Cell Derived Factor-1 alpha/CXCL12 (CHO-expressed) is a member of CXC chemokine family, binds to CXCR4 and CXCR7, and is implicated in tumor proliferation, survival, vascularization, metastasis, and immunosuppression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3160 Supplier Medchemexpress LLC Supplier No. HYP740310UG
Medchemexpress, HY-P7403 10ug SDF-1α/CXCL12 (CHO-expressed), Mouse CAS: Recombinant Mouse Stromal-Cell Derived Factor-1 alpha/CXCL12 (CHO-expressed) is a member of CXC chemokine family, binds to CXCR4 and CXCR7, and is implicated in tumor proliferation, survival, vascularization, metastasis, and immunosuppression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.