Learn More
Sino Biological Recombinant Human SDF-1/CXCL12 Protein
Supplier:  Sino Biological 10118HNAE5
Encompass_Preferred
A DNA sequence encoding the human CXCL12 isoform beta (P48061-1) (Lys 22-Met 93) was expressed and purified
Catalog No. 50-211-4920