Learn More
Aviva Systems Biology Corp SDF2 Antibody - N-terminal region (ARP48718_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48718_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SDF2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SDF2. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59897