Learn More
Aviva Systems Biology Corp SDF4 Antibody - C-terminal region (ARP47222_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47222_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SDF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SDF4. Alternate names: Cab45, RP5-902P8.6, SDF-4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-59490