Learn More
Aviva Systems Biology Corp Sdhb Antibody - middle region (ARP47980_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47980_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sdhb. It was validated on Western Blot. . Alternate names: 0710008N11RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-337