Learn More
Aviva Systems Biology Corp SDK1 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP45025_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SDK1. It was validated on Western Blot. SDK1,sidekick homolog 1, cell adhesion molecule (chicken),FLJ31425; SDK1,protein sidekick-1,protein sidekick-1,OTTHUMP00000200448The immunogen is a synthetic peptide directed towards the middle region of human SDK1. Alternate names: FLJ31425ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Zebrafish
Catalog No. 50-285-177