Learn More
Medchemexpress LLC SDMA (Synonyms: Symmetric dimethylarginine; NG,NG'-Dimethyl-L-arginine) 5mg
SDP
Catalog No. NC3242640
Encompass_Preferred
SDMA (Symmetric dimethylarginine) is an endogenous inhibitor of nitric oxide (NO) synthase activity. SDMA, a novel kidney biomarker, permits earlier diagnosis of kidney disease than traditional creatinine testing.
Catalog No. NC3242640 Supplier Medchemexpress LLC Supplier No. HY1014105MG
SDMA (Symmetric dimethylarginine) is an endogenous inhibitor of nitric oxide (NO) synthase activity. SDMA, a novel kidney biomarker, permits earlier diagnosis of kidney disease than traditional creatinine testing.